MilliporeSigma
  • Synchronous circadian voltage rhythms with asynchronous calcium rhythms in the suprachiasmatic nucleus.
Synchronous circadian voltage rhythms with asynchronous calcium rhythms in the suprachiasmatic nucleus.
Proceedings of the National Academy of Sciences of the United States of America (2017-03-09)
Ryosuke Enoki, Yoshiaki Oda, Michihiro Mieda, Daisuke Ono, Sato Honma, Ken-Ichi Honma
ABSTRACT
The suprachiasmatic nucleus (SCN), the master circadian clock, contains a network composed of multiple types of neurons which are thought to form a hierarchical and multioscillator system. The molecular clock machinery in SCN neurons drives membrane excitability and sends time cue signals to various brain regions and peripheral organs. However, how and at what time of the day these neurons transmit output signals remain largely unknown. Here, we successfully visualized circadian voltage rhythms optically for many days using a genetically encoded voltage sensor, ArcLightD. Unexpectedly, the voltage rhythms are synchronized across the entire SCN network of cultured slices, whereas simultaneously recorded Ca