MilliporeSigma
  • Lovastatin upregulates microRNA-29b to reduce oxidative stress in rats with multiple cardiovascular risk factors.
Lovastatin upregulates microRNA-29b to reduce oxidative stress in rats with multiple cardiovascular risk factors.
Oncotarget (2017-01-07)
Fu Wang, Hui Ma, Wen-Jing Liang, Jing-Jing Yang, Xue-Qing Wang, Mei-Rong Shan, Yuan Chen, Min Jia, Ya-Ling Yin, Xue-Ying Sun, Jia-Ning Zhang, Qi-Sheng Peng, Yu-Guo Chen, Li-Ying Liu, Peng Li, Tao Guo, Shuang-Xi Wang
ABSTRACT
Proteasome-linked oxidative stress is believed to cause endothelial dysfunction, an early event in cardiovascular diseases (CVD). Statin, as HMG-CoA reductase inhibitor, prevents endothelial dysfunction in CVD. However, the molecular mechanism of statin-mediated normalization of endothelial function is not completely elucidated. Lovastatin time/dose-dependently increased miR-29b expression and decreased proteasome activity in cultured human umbilical vein endothelial cells (HUVECs). Anti-miR-29b or overexpression of PA200 abolished lovastatin-induced inhibition of proteasome activity in HUVECs. In contrast, pre-miR-29b or PA200 siRNA mimics these effects of lovastatin on proteasome activity. Lovastatin inhibited oxidative stress induced by multiple oxidants including ox-LDL, H2O2, TNFα, homocysteine thiolactone (HTL), and high glucose (HG), which were reversed by inhibition of miR-29b in HUVECs. Ex vivo analysis indicated that lovastatin normalized the acetylcholine-induced endothelium-dependent relaxation and the redox status in isolated rat aortic arteries exposure to multiple cardiovascular risk factors. In vivo analysis revealed that administration of lovastatin remarkably suppressed oxidative stress and prevented endothelial dysfunction in rats with hyperglycemia, dyslipidemia, and hyperhomocysteinemia, as well as increased miR-29b expressions, reduced PA200 protein levels, and suppression of proteasome activity in aortic tissues. Upregulation of miR-29b expression is a common mechanism contributing to endothelial dysfunction induced by multiple cardiovascular risk factors through PA200-dependent proteasome-mediated oxidative stress, which is prevented by lovastatin.