MilliporeSigma
  • Prevalence of vesivirus in a laboratory-based set of serum samples obtained from dairy and beef cattle.
Prevalence of vesivirus in a laboratory-based set of serum samples obtained from dairy and beef cattle.
American journal of veterinary research (2006-01-24)
Andreas Kurth, James F Evermann, Douglas E Skilling, David O Matson, Alvin W Smith
ABSTRACT
To examine sera obtained from dairy and beef cattle to detect antibodies against vesivirus and compare seroprevalence among cattle within the sample population. Cattle sera from 8 western states and Maryland submitted to the Washington Animal Disease Diagnostic Laboratory during 1999 and 2000. Sera were analyzed for vesivirus-specific antibodies by use of a recombinant vesivirus-San Miguel sea lion virus serotype 5-capsid peptide antigen in an indirect ELISA. Overall, 693 sera were tested and 105 (15.2%) had positive results. Seropositive cattle were from 7 states (all cattle from Montana and Maryland 10 and 4, respectively were seronegative). Overall seroprevalence for antivesivirus antibody in herds ranged between 0% and 80% (median, 14%). Higher antibody prevalence was significantly associated with older age, dairy rather than beef cattle, and reasons for submission. Logistic regression of factors (abortion, respiratory tract disease, and all other reasons for sample submission) revealed that older age and other reasons were independently associated with higher seroprevalence. Higher seropositive optical density values for the ELISA were observed among older cattle and cattle that aborted, compared with values for cattle with respiratory tract disease or other reasons for submission. This laboratory-based surveillance sample provided a point estimate of seroprevalence against vesivirus among cattle in 9 US states. This suggests that vesivirus infection is widespread with high prevalence in some herds. Risk factors associated with vesivirus seroprevalence in beef and dairy cattle should be confirmed in population-based studies.