MilliporeSigma
  • A comparison of three methods of soft ionization mass spectrometry of crude phospholipid extracts.
A comparison of three methods of soft ionization mass spectrometry of crude phospholipid extracts.
Biomedical mass spectrometry (1985-11-01)
E Benfenati, R Reginato
ABSTRACT
Many different classes of phospholipids were identified from crude extracts of hearts by three soft ionization mass spectrometric techniques: liquid matrix secondary ion mass spectrometry in the negative mode, (-)LSIMS, and in the positive mode, (+)LSIMS, and field desorption. (-)LSIMS and (+)LSIMS are complementary methods. In some cases it was possible to establish the fatty acid and aldehyde composition and position of some phospholipid classes, by the analysis of fragments.