MilliporeSigma
  • Regulatory cytokine production stimulated by DNA vaccination against an altered form of glutamic acid decarboxylase 65 in nonobese diabetic mice.
Regulatory cytokine production stimulated by DNA vaccination against an altered form of glutamic acid decarboxylase 65 in nonobese diabetic mice.
Journal of molecular medicine (Berlin, Germany) (2003-04-12)
Yelena Glinka, Renée De Pooter, France Croze, Gérald J Prud'homme
ABSTRACT
Nonobese diabetic (NOD) mice develop a T-cell dependent autoimmune form of diabetes, in which glutamic acid decarboxylase 65 (GAD65) is an important islet target antigen. Intramuscular DNA vaccination with a plasmid encoding native GAD65 (a cytosolic antigen) did not significantly alter the incidence of diabetes, but vaccination against an altered form of GAD65 with a signal peptide (spGAD), which is secreted in vitro, was protective. The preventive effect was further enhanced by repeated injections of the spGAD plasmid. Following DNA injection into muscle GAD65 was expressed for several months, and this was not accompanied by an inflammatory response. Immunization against GAD65 was not associated with substantial alterations in cytokine production by splenic lymphocytes stimulated with immunogenic GAD65 peptides. In contrast, spGAD induced increased secretion of both interleukin 10 and interferon gamma and a striking decrease in the interferon gamma/interleukin 10 ratio in culture supernatants. Similarly, spGAD-immunized mice had higher serum interleukin 10 levels and lower serum interferon gamma levels than other groups, suggesting a systemic effect. In nondiabetic mice there was increased basal production of transforming growth factor beta(1), which was enhanced by antigenic stimulation. These alterations in regulatory cytokine production were apparent both early and late after the treatment was initiated. These findings suggest that DNA vaccination against spGAD protects NOD mice by increasing regulatory cytokine production.