MilliporeSigma
  • Calponin expression in laryngeal myoepithelial carcinoma and its prognostic implications: a case report and literature review.
Calponin expression in laryngeal myoepithelial carcinoma and its prognostic implications: a case report and literature review.
The Journal of international medical research (2010-06-03)
Y J Mao, X M Luo, S H Zhou, Z J Zheng
ABSTRACT
A case report of laryngeal myoepithelial carcinoma (MEC) is presented and the literature concerning prognostic factors in MEC is reviewed. A 61-year old man was admitted to hospital with hoarseness and progressive dyspnoea. On examination, both vocal cords were fixed in the midline with a glottic fissure of only 3 mm. No tumour was seen, but the subglottis was not completely visible. A computed tomography scan showed a soft mass below the right vocal cord obstructing two-thirds of the larynx. On suspension laryngoscopy, a dull mass (1.5 x 1.5 cm) was seen below the right vocal cord, which was malignant on frozen biopsy. A total laryngectomy was performed and the patient received radiotherapy. He died of recurrence 25 months later. The tumour was positive for cytokeratin 14, S-100 protein and calponin. MEC of the larynx is extremely rare. The clinical behaviour of MEC is variable and prognostic factors have been poorly analysed. Calponin expression may be a prognostic factor, but other factors also affect the outcome in MEC.