MilliporeSigma
  • Genome-wide repression of eRNA and target gene loci by the ETV6-RUNX1 fusion in acute leukemia.
Genome-wide repression of eRNA and target gene loci by the ETV6-RUNX1 fusion in acute leukemia.
Genome research (2016-11-03)
Susanna Teppo, Saara Laukkanen, Thomas Liuksiala, Jessica Nordlund, Mikko Oittinen, Kaisa Teittinen, Toni Grönroos, Pascal St-Onge, Daniel Sinnett, Ann-Christine Syvänen, Matti Nykter, Keijo Viiri, Merja Heinäniemi, Olli Lohi
ABSTRACT
Approximately 20%-25% of childhood acute lymphoblastic leukemias carry the ETV6-RUNX1 (E/R) fusion gene, a fusion of two central hematopoietic transcription factors, ETV6 (TEL) and RUNX1 (AML1). Despite its prevalence, the exact genomic targets of E/R have remained elusive. We evaluated gene loci and enhancers targeted by E/R genome-wide in precursor B acute leukemia cells using global run-on sequencing (GRO-seq). We show that expression of the E/R fusion leads to widespread repression of RUNX1 motif-containing enhancers at its target gene loci. Moreover, multiple super-enhancers from the CD19