MilliporeSigma
  • Tissue-specific differences in heat shock protein hsc70 and hsp70 in the control and hyperthermic rabbit.
Tissue-specific differences in heat shock protein hsc70 and hsp70 in the control and hyperthermic rabbit.
Journal of cellular physiology (1997-02-01)
P Manzerra, S J Rush, I R Brown
ABSTRACT
The ability to resolve protein members of the hsp70 multigene family by two-dimensional Western blotting permitted the characterization of antibodies which were specific in discriminating constitutively expressed hsc70 isoforms from stress-inducible hsp70 isoforms. This antibody characterization demonstrated that basal levels of hsp70 isoforms were present in the cerebellum of the control rabbit and that these were elevated following hyperthermia, whereas levels of hsc70 were similar in control and hyperthermic tissue. Multiple isoforms of hsp70 were detected but tissue-specific differences were not apparent in various organs of the rabbit. However, species differences were observed as fewer hsp70 isoforms were noted in rat and mouse. In the control rabbit, higher levels of hsc70 protein were present in neural tissues compared to non-neural tissues. Following physiologically relevant hyperthermia, induction of hsp70 was greatest in non-neural tissues such as liver, heart, muscle, spleen, and kidney compared to regions of the nervous system. These studies suggest that the amount of preexisting constitutive hsc70 protein may influence the level of induction of hsp70 in the stress response. Given this observation, caution is required in the employment of hsp70 induction as an index of cellular stress since endogenous levels of hsc70, and perhaps hsp70, may modulate the level of induction.