MilliporeSigma
  • Identification of 20 genes aberrantly methylated in human breast cancers.
Identification of 20 genes aberrantly methylated in human breast cancers.
International journal of cancer (2005-04-09)
Kazuaki Miyamoto, Takashi Fukutomi, Sadako Akashi-Tanaka, Tadashi Hasegawa, Toshimasa Asahara, Takashi Sugimura, Toshikazu Ushijima
ABSTRACT
Aberrant methylation of CpG islands (CGI) not only plays a role in gene silencing, but is also a potential cancer biomarker. To identify more CGI aberrantly methylated in human breast cancers, we carried out a genome-wide search for aberrant methylation, using methylation-sensitive-representational difference analysis. CGI in 5' upstream regions of 20 genes, TSPAN-2, AK5, LOC284999, HOXD11, FLJ25161, XT3, PCDH10, PCDHGB6, SIM1, LOC346978, COE2, TDH (FLJ25033), LOC346419, FLJ33790, GJB2, AMN, LOC201164, DLX4, DCC and FOXA2, were found to be methylated in at least one of 8 breast cancer cell lines. Fifteen of the 20 genes were methylated in more than one of 21 primary breast cancers in Stages I or II, and especially, those of LOC346978, HOXD11, SIM1, PCDHGB6 and FLJ25161 were methylated in more than 10 cancers. All the breast cancers had some aberrant methylation. Among the 13 genes whose CGI were completely methylated in one or more cell lines, FOXA2 and XT3 were expressed in normal human mammary epithelial cells (HMEC) and were not expressed in cancer cell lines with complete methylation. The other 11 genes examined were barely expressed, or not expressed even in HMEC. Our results showed that breast cancer cells accumulate aberrant methylation of the CGI identified here. This may serve as markers for early-stage breast cancers and suggests that aberrant methylation targets transcriptionally inactive genes in vivo.