MilliporeSigma
  • SATB1 is an independent prognostic factor in radically resected upper gastrointestinal tract adenocarcinoma.
SATB1 is an independent prognostic factor in radically resected upper gastrointestinal tract adenocarcinoma.
Virchows Archiv : an international journal of pathology (2014-10-20)
Charlotta Hedner, Alexander Gaber, Dejan Korkocic, Björn Nodin, Mathias Uhlén, Eugenia Kuteeva, Henrik Johannesson, Karin Jirström, Jakob Eberhard
ABSTRACT
Gastric cancer is the second most common cause of cancer-related death worldwide, and the incidence of esophageal adenocarcinoma is rising. While some progress has been made in treatment strategies, overall survival remains very poor for patients with adenocarcinoma in the upper gastrointestinal tract. Special AT-rich sequence binding protein 1 (SATB1) is a global genome organizer that has been demonstrated to promote aggressive tumor behavior in several different types of cancer, including gastric cancer. The prognostic value of SATB1 expression in esophageal cancer has, however, not yet been described. In this study, expression of SATB1 was examined by immunohistochemistry on tissue microarrays prepared from tissue samples from 175 patients with adenocarcinoma of the esophagus, cardia, or stomach and containing normal tissue, intestinal metaplasia, primary tumors, and metastases. A well-validated antibody was used. We found SATB1 to be an independent prognostic factor in patients with a radically resected tumor, correlating with shorter overall survival as well as with shorter recurrence-free survival. SATB1 expression was also found to be significantly lower in primary tumors associated with intestinal metaplasia than those without intestinal metaplasia. This observation is of potential biological interest as it has been proposed that intestinal metaplasia-associated tumors constitute a less aggressive phenotype.