MilliporeSigma
  • Syndecan-1 knockdown in endometrial epithelial cells alters their apoptotic protein profile and enhances the inducibility of apoptosis.
Syndecan-1 knockdown in endometrial epithelial cells alters their apoptotic protein profile and enhances the inducibility of apoptosis.
Molecular human reproduction (2014-01-31)
S J Boeddeker, D M Baston-Buest, O Altergot-Ahmad, J S Kruessel, A P Hess
ABSTRACT
Endometrial epithelial cells are known to undergo apoptosis during trophoblast invasion. We postulate that the cell surface molecule Syndecan-1 which is expressed on endometrial cells and syncytiotrophoblast is important for implantation in general and especially for induction of maternal cell apoptosis during trophoblast invasion because Syndecan-1's influence on apoptotic susceptibility of cancer cells is already described in the literature. Using the human endometrial epithelial cell line RL95-2, a new stable cell line with Syndecan-1 knockdown was generated. Via antibody array analysis, a significant decrease in the expression of anti-apoptotic proteins like inhibitors of apoptosis, Clusterin, heme oxygenase (HO-2), heat shock protein (HSP)27 and -70 and Survivin due to the Syndecan-1 knockdown was discovered. Correspondingly, active Caspase-3 as an indicator for apoptosis was increased more severely in these cells compared with unmodified RL95-2 after treatment with implantation-related stimuli, which are the cytokines interleukin-1β, interferon-γ, tumor necrosis factor-α and transforming growth factor-β1 and an anti-Fas antibody. Furthermore, a treatment with a combination of all factors caused a higher Caspase-3 induction compared with each single treatment. These results demonstrate that Syndecan-1 is involved in the control of apoptosis in RL95-2 cells and therefore may affect the fine tuning of apoptosis in endometrial epithelium regulating the embryo's invasion depth as a crucial step for regular implantation followed by successful pregnancy.