MilliporeSigma
  • Fusion of Golgi-derived vesicles mediated by SNAP-25 is essential for sympathetic neuron outgrowth but relatively insensitive to botulinum neurotoxins in vitro.
Fusion of Golgi-derived vesicles mediated by SNAP-25 is essential for sympathetic neuron outgrowth but relatively insensitive to botulinum neurotoxins in vitro.
The FEBS journal (2014-05-28)
Gary W Lawrence, Jiafu Wang, Mitchell F Brin, K Roger Aoki, Larry Wheeler, J Oliver Dolly
ABSTRACT
Sympathetic neurons ramify to innervate multiple cells in target tissues. In compartmentalized cultures of rat superior cervical ganglion neurons, cleavage of synaptosomal-associated protein of Mr  = 25 000 (SNAP-25) in neurites exposed to botulinum neurotoxin type A (BoNT/A) arrested their growth and collapsed interstitial branches, but this required large, nonclinical doses. A protease-inactive mutant proved ineffective, confirming involvement of SNAP-25 in neurite extension and arborization. BoNT/C1 acted like BoNT/A, but BoNT/E caused only mild inhibition, likely due to transient SNAP-25 proteolysis. Near-total lack of susceptibility to BoNT/B or BoNT/D revealed that vesicle-associated membrane protein (VAMPs) isoforms 1-3 are not essential. Neurite length was not reduced when either BoNT/A or BoNT/C1 was applied to the somata, with no detrimental effect on neuron viability being observed. Treatments that protect cells from deprivation of nerve growth factor failed to prevent the toxin-induced loss of neurites. Inactivation of SNAP-25 caused the accumulation at neurite branch sites of Golgi-derived organelles labelled with N-(4,4-difluoro-5,7-dimethyl-4-bora-3a,4a-diaza-s-indacene-3-pentanoyl)-sphingosine conjugated to bovine serum albumin, prior to the collapse of arbors. Notably, neurite growth was ~ 1000-fold less susceptible to BoNT/A than cholinergic transmission in these neurons. Accordingly, a BoNT/A acceptor synaptic vesicle protein 2 (SV2) was found to be colocalized with VAMP 1-3, but not with VAMP 7, which is implicated in the growth of neurites. In conclusion, neurites depend on SNAP-25 for extension but this is quite resistant to BoNT/A, possibly, because of a low density of SV2 at growth sites that are distant from the highly susceptible regions of neurotransmitter release.