MilliporeSigma
  • Cerebellar stem cells do not produce neurons and astrocytes in adult mouse.
Cerebellar stem cells do not produce neurons and astrocytes in adult mouse.
Biochemical and biophysical research communications (2014-06-20)
Xin Su, Wuqiang Guan, Yong-Chun Yu, Yinghui Fu
ABSTRACT
Although previous studies implied that cerebellar stem cells exist in some adult mammals, little is known about whether these stem cells can produce new neurons and astrocytes. In this study by bromodeoxyuridine (BrdU) intraperitoneal (i.p.) injection, we found that there are abundant BrdU(+) cells in adult mouse cerebellum, and their quantity and density decreases significantly over time. We also found cell proliferation rate is diversified in different cerebellar regions. Among these BrdU(+) cells, very few are mash1(+) or nestin(+) stem cells, and the vast majority of cerebellar stem cells are quiescent. Data obtained by in vivo retrovirus injection indicate that stem cells do not produce neurons and astrocytes in adult mouse cerebellum. Instead, some cells labeled by retrovirus are Iba1(+) microglia. These results indicate that very few stem cells exist in adult mouse cerebellum, and none of these stem cells contribute to neurogenesis and astrogenesis under physiological condition.