MilliporeSigma
  • Loss of Vhl in cartilage accelerated the progression of age-associated and surgically induced murine osteoarthritis.
Loss of Vhl in cartilage accelerated the progression of age-associated and surgically induced murine osteoarthritis.
Osteoarthritis and cartilage (2014-07-08)
T Weng, Y Xie, L Yi, J Huang, F Luo, X Du, L Chen, C Liu, D Chen, L Chen
ABSTRACT
To investigate the role of Vhl in maintaining the integrity of articular cartilage and in the development of experimental osteoarthritis (OA). Histology of articular cartilage and subchondral bone in both Vhl cKO and WT mice were analyzed by histopathology and micro-CT. Articular cartilage destruction and proteoglycan loss were scored in aged (12-month-old) mice as well as in mice with surgically induced OA. Apoptosis of cartilage in age-related and surgically induced OA was detected with TUNEL assay. Expressions of von Hippel-Lindau (VHL), Fas, LC-3, HIF-1α, HIF-2α, p-mTOR and MMP-13 in the knee joints were analyzed by immunostaining. No gross differences in cartilage were observed between Vhl cKO and WT mice at age 4 months. However, Vhl cKO mice displayed accelerated age-associated spontaneous OA and surgically induced OA. Cartilage destruction and proteoglycan loss were observed in the absence of Vhl. In addition, inactivation of Vhl resulted in up-regulation of HIF-2α and increased chondrocyte apoptosis and decreased expression of autophagy during OA development. Immunohistochemical staining also showed that Vhl deficiency led to increased expression of Fas, p-mTOR and MMP-13, and those genes were associated with cell apoptosis, autophagy and cartilage matrix breakdown, respectively. Loss of Vhl in adult articular cartilage is associated with earlier dysregulation of cartilage homeostasis, characterized by an increased chondrocyte apoptosis, compromised chondrocyte autophagy, and an accelerated age-related and surgery-induced OA development. These results highlight the novel role of Vhl in maintaining joint homeostasis and OA development.