MilliporeSigma
  • Phosphorylated insulin-like growth factor-1 receptor expression predicts poor prognosis of Chinese patients with gastric cancer.
Phosphorylated insulin-like growth factor-1 receptor expression predicts poor prognosis of Chinese patients with gastric cancer.
Medical oncology (Northwood, London, England) (2014-10-29)
Wen-ying Deng, Ning Li, Xiang-bin Wan, Su-xia Luo, You-wei Zhang
ABSTRACT
Previous studies have established the role of phosphorylated form of insulin-like growth factor type 1 receptor (p-IGF1R) as a good candidate for tumor biomarker. The aims of this study were to investigate p-IGF1R expression status in gastric cancer (GC) specimens and to clarify its clinical significance. A total of 78 GC patients treated with radical resection were enrolled in this study. Immunohistochemistry was used to detect p-IGF1R and phosphatase and tensin homolog deleted on chromosome ten (PTEN) protein expression in paired tumor and adjacent normal tissues. Results showed a higher level of p-IGF1R protein expression in tumor tissues than that in normal tissues, and the rate of p-IGF1R protein high/moderate expression in GC and normal tissues was 52.6% (41/78) and 6.4% (5/78), respectively (p < 0.001). In contrast, PTEN protein expression was downregulated in GC, as compared with normal tissues (negative/low expression 49/78 vs. 8/78, p < 0.001). Moreover, PTEN protein downregulation was consistent with p-IGF1R upregulation. Overexpression of p-IGF1R protein was associated with lymph metastasis, clinical stage, and adverse 3-year progression-free survival (PFS). Survival analysis and Cox proportional hazards model revealed that p-IGF1R overexpression was an independent factor in predicting PFS for GC patients, apart from lymph metastasis. In conclusion, p-IGF1R is highly expressed in GC, which may be a novel biomarker to predict the clinical outcome of GC patients.