MilliporeSigma
  • Characterization of a branched-chain amino-acid transporter SBAT1 (SLC6A15) that is expressed in human brain.
Characterization of a branched-chain amino-acid transporter SBAT1 (SLC6A15) that is expressed in human brain.
Biochemical and biophysical research communications (2005-10-18)
Hitomi Takanaga, Bryan Mackenzie, Ji-Bin Peng, Matthias A Hediger
ABSTRACT
The SLC6 gene family comprises membrane proteins that transport neurotransmitters, amino acids, or osmolytes. We report the first functional characterization of the human SLC6A15 gene, which codes for a sodium-coupled branched-chain amino-acid transporter 1 (SBAT1). SBAT1 expression is specific to the brain. When expressed in Xenopus oocytes, SBAT1 mediated Na+-coupled transport of hydrophobic, zwitterionic alpha-amino and imino acids. SBAT1 exhibited a strong preference for branched-chain amino acids (BCAA) and methionine (K0.5 80-160 microM). SBAT1 excluded aromatic or charged amino acids, beta-amino acids, glycine, and GABA. SBAT1-mediated transport of amino or imino acids was extremely temperature-dependent (Q10=9) and was inhibited at acidic pH. PKC activation reduced the plasma-membrane population of SBAT1 protein. SBAT1-mediated transport of BCAA, particularly leucine, may be an important source of amino nitrogen for neurotransmitter synthesis in glutamatergic and GABAergic neurons.