MilliporeSigma
  • Ubiquitin-specific peptidase 20 targets TRAF6 and human T cell leukemia virus type 1 tax to negatively regulate NF-kappaB signaling.
Ubiquitin-specific peptidase 20 targets TRAF6 and human T cell leukemia virus type 1 tax to negatively regulate NF-kappaB signaling.
Journal of virology (2011-04-29)
Junichiro Yasunaga, Frank C Lin, Xiongbin Lu, Kuan-Teh Jeang
ABSTRACT
NF-κB plays a key role in innate and acquired immunity. Its activity is regulated through intricate signaling networks. Persistent or excessive activation of NF-κB induces diseases, such as autoimmune disorders and malignant neoplasms. Infection by human T cell leukemia virus type 1 (HTLV-1) causes a fatal hematopoietic malignancy termed adult T cell leukemia (ATL). The HTLV-1 viral oncoprotein Tax functions pivotally in leukemogenesis through its potent activation of NF-κB. Recent findings suggest that protein ubiquitination is crucial for proper regulation of NF-κB signaling and for Tax activity. Here, we report that ubiquitin-specific peptidase USP20 deubiquitinates TRAF6 and Tax and suppresses interleukin 1β (IL-1β)- and Tax-induced NF-κB activation. Our results point to USP20 as a key negative regulator of Tax-induced NF-κB signaling.