MilliporeSigma
  • Downregulation of robust acute type I interferon responses distinguishes nonpathogenic simian immunodeficiency virus (SIV) infection of natural hosts from pathogenic SIV infection of rhesus macaques.
Downregulation of robust acute type I interferon responses distinguishes nonpathogenic simian immunodeficiency virus (SIV) infection of natural hosts from pathogenic SIV infection of rhesus macaques.
Journal of virology (2010-05-21)
Levelle D Harris, Brian Tabb, Donald L Sodora, Mirko Paiardini, Nichole R Klatt, Daniel C Douek, Guido Silvestri, Michaela Müller-Trutwin, Ivona Vasile-Pandrea, Cristian Apetrei, Vanessa Hirsch, Jeffrey Lifson, Jason M Brenchley, Jacob D Estes
ABSTRACT
The mechanisms underlying the AIDS resistance of natural hosts for simian immunodeficiency virus (SIV) remain unknown. Recently, it was proposed that natural SIV hosts avoid disease because their plasmacytoid dendritic cells (pDCs) are intrinsically unable to produce alpha interferon (IFN-alpha) in response to SIV RNA stimulation. However, here we show that (i) acute SIV infections of natural hosts are associated with a rapid and robust type I IFN response in vivo, (ii) pDCs are the principal in vivo producers of IFN-alpha/beta at peak acute infection in lymphatic tissues, and (iii) natural SIV hosts downregulate these responses in early chronic infection. In contrast, persistently high type I IFN responses are observed during pathogenic SIV infection of rhesus macaques.