MilliporeSigma
[Bufexamac-induced pigmented purpuric eruption].
Der Hautarzt; Zeitschrift fur Dermatologie, Venerologie, und verwandte Gebiete (2008-12-19)
K Waltermann, W Ch Marsch, B Kreft
ABSTRACT
We report on a case of a bufexamac-induced allergic contact dermatitis with hematogenous dissemination presenting with the clinical and histological picture of a pigmented purpuric eruption. To our knowledge this is the first report on a bufexamac-induced pigmented purpuric dermatosis. It represents a further example of the clinical variety of cutaneous side-effects caused by bufexamac.