MilliporeSigma
Paeoniflorin acts as a liver X receptor agonist.
Journal of Asian natural products research (2013-01-04)
Hsiang-Ru Lin
ABSTRACT
Paeoniflorin is one of the active ingredients of Paeonia lactiflora Pall., a novel traditional herbal medicine exerting pharmacological effects including antihyperlipidemic, neuroprotective, and anti-hepatofibrosis effects. Liver X receptor (LXR) acts as a ligand-activated transcription factor to exhibit antihyperlipidemic and neuroprotective effects. In this study, the activity of paeoniflorin against LXR was evaluated by the mammalian one-hybrid and transient transfection reporter assays. The results showed that paeoniflorin transactivated GAL4, rat cholesterol 7 α-hydroxylase, phospholipid transfer protein, and ATP-binding cassette A1 gene promoters in dose-dependent manner. Furthermore, the docking study demonstrated that paeoniflorin resided in the LXR ligand-binding pocket in the similar manner as GSK 3987, a novel LXR agonist. These results indicated that paeoniflorin might exert pharmacological effects through LXR pathway.
Sigma-Aldrich
Paeoniflorin, ≥98% (HPLC)