MilliporeSigma
  • Thyroidectomy followed by fosbretabulin (CA4P) combination regimen appears to suggest improvement in patient survival in anaplastic thyroid cancer.
Thyroidectomy followed by fosbretabulin (CA4P) combination regimen appears to suggest improvement in patient survival in anaplastic thyroid cancer.
Surgery (2012-11-20)
Julie A Sosa, Jai Balkissoon, Shiao-ping Lu, Peter Langecker, Rossella Elisei, Barbara Jarzab, C S Bal, Shanthi Marur, Ann Gramza, Frank Ondrey
ABSTRACT
Anaplastic thyroid cancer (ATC) is an aggressive neoplasm for which a paucity of data exist about the relative role of operative procedures in disease management. The FACT trial was a randomized, controlled phase 2/3 trial assessing the safety and efficacy of carboplatin/paclitaxel with CA4P (experimental arm) or without CA4P (control arm) in ATC, 2007-11. Patients were permitted to have had an operation before enrollment, which was stratified on the basis of exposure to operation. A subpopulation of patients who had a cancer-related operation (thyroidectomy) was compared with those who did not, and 1-year and median survival were estimated. A total of 80 patients were enrolled; 55% had undergone a cancer-related operation, of whom 70% had near-total/total thyroidectomy. Baseline characteristics for operative and nonoperative patients were not substantially different. Median survival for patients who had cancer-related operation was 8.2 months in the CA4P arm versus 4.0 months in the control arm, resulting in a hazard ratio of 0.66 (P = .25) and a suggested associated reduction in risk of death of 35%. 1-year survival was 33.3% in the CA4P arm versus 7.7% in the control arm. In this largest prospective study ever conducted in ATC, thyroidectomy followed by CA4P combination regimen showed a nonsignificant trend toward improvement in patient survival.
Sigma-Aldrich
Combretastatin A4, ≥98% (HPLC), powder