MilliporeSigma
  • Metallophilic bonding and agostic interactions in gold(I) and silver(I) complexes bearing a thiotetrazole unit.
Metallophilic bonding and agostic interactions in gold(I) and silver(I) complexes bearing a thiotetrazole unit.
Inorganic chemistry (2011-02-12)
Adriana Ilie, Ciprian I Raţ, Susanne Scheutzow, Christiane Kiske, Karin Lux, Thomas M Klapötke, Cristian Silvestru, Konstantin Karaghiosoff
ABSTRACT
Gold(I) and silver(I) complexes of 1-methyl-5-thio-tetrazole (1) have been prepared and the coordination chemistry of this ligand toward metal-phosphine frameworks has been explored. As indicated by IR and Raman data, ligand 1 is deprotonated and the resulted anion acts as a bidentate (S,N)-tetrazole-5-thiolato unit in the new gold(I) complexes, [Au(SCN(4)Me)(PPh(3))] (2), [{Au(SCN(4)Me)}(2)(μ-dppm)] (3), and [{Au(SCN(4)Me)}(2)(μ-dppe)] (4), while it is coordinated only through the sulfur atom as its neutral tetrazole-5-thione form in the silver(I) derivative, [Ag(HSCN(4)Me)(PPh(3))](2)(OTf)(2) (5). Further characterization of the new compounds was performed using multinuclear ((1)H, (13)C, (31)P, (19)F) NMR spectroscopy, mass spectrometry, and DSC measurements. Single-crystal X-ray diffraction studies revealed basically linear P-M-S arrangements in complexes 3-5. The bidentate (S,N) coordination pattern results in a T-shaped (S,N)PAu core in 3 and 4, whereas, in 5, a similar coordination geometry is achieved in the dimer association based on S-bridging ligand 1. Herein, weak (C)H···Au and (C)H···Ag agostic interactions were observed. An intramolecular Au···Au contact occurs in 3, while in 4 intermolecular aurophilic bonds lead to formation of a chain polymer. An intermolecular Ag···Ag contact is also present in the dimer unit of 5. Low-temperature (31)P NMR data for 5 evidenced the presence of monomer and dimer units in solution. Theoretical calculations on model of the complexes 2 and 4 are consistent with the geometries found by X-ray diffraction studies.