MilliporeSigma
Effect of cholesterol on diffusion in surfactant bilayers.
The Journal of chemical physics (2007-11-06)
Thorsten Pieper, Svetlana Markova, Masataka Kinjo, Dieter Suter
ABSTRACT
Biological membranes consist of lipid bilayers with liquid-ordered and liquid-disordered phases. It is believed that cholesterol controls the size of the microdomains in the liquid-ordered phase and thereby affects the mobility as well as the permeability of the membrane. We study this process in a model system consisting of the nonionic surfactant C(12)E(5) and water in the lamellar phase. We measure the diffusion of fluorescent probe molecules (rhodamine B) by fluorescence correlation spectroscopy. For different surfactant to water ratios, we measure how the molecular mobility varies with the amount of cholesterol added. We find that a reduction of the diffusion coefficient is already detectable at a molar ratio of 8 mol % cholesterol.
Sigma-Aldrich
Pentaethylene glycol monododecyl ether, BioXtra, ≥98.0% (GC)