MilliporeSigma
Antiplaque and antigingivitis effectiveness of a hexetidine mouthwash.
Journal of clinical periodontology (2003-07-02)
N C Sharma, H J Galustians, J Qaqish, C H Charles, J W Vincent, J A McGuire
ABSTRACT
To assess the antiplaque/antigingivitis efficacy of a hexetidine-containing mouthwash. This examiner-blind, parallel group, controlled clinical study examined the effectiveness of a hexetidine (0.1%) mouthwash both in inhibiting the development of supragingival plaque and in reducing gingivitis. One hundred and thirty-four adult subjects completed the 2-week experimental gingivitis model study. Following baseline examinations, which included plaque index, modified gingival index and gingival bleeding index, subjects received a full dental prophylaxis. Subjects were randomly assigned to one of three mouthwashes (hexetidine 0.1%, chlorhexidine 0.12% (positive control) or a 5% hydroalcohol negative control) and commenced three times daily supervised rinsing as their sole method of oral hygiene. All indices were rescored after 2 weeks. Compared to the negative control group, the hexetidine group demonstrated a statistically significant inhibition and reduction of supragingival plaque and gingival inflammation with reductions of 6.3%, 33.5% and 56% for gingivitis, plaque and gingival bleeding, respectively. The results of the chlorhexidine group were used to validate the study. The study confirms the efficacy of a hexetidine rinse in reducing supragingival plaque and gingival inflammation.