MilliporeSigma
  • The role of peripheral 5-HT1A, 5-HT1B, 5-HT1D, 5-HT1E and 5-HT1F serotonergic receptors in the reduction of nociception in rats.
The role of peripheral 5-HT1A, 5-HT1B, 5-HT1D, 5-HT1E and 5-HT1F serotonergic receptors in the reduction of nociception in rats.
Neuroscience (2009-10-20)
V Granados-Soto, C F Argüelles, H I Rocha-González, B Godínez-Chaparro, F J Flores-Murrieta, C M Villalón
ABSTRACT
This study assessed the possible antinociceptive role of peripheral 5-HT(1) receptor subtypes in the rat formalin test. Rats were injected into the dorsum of the hind paw with 50 microl of diluted formalin (1%). Nociceptive behavior was quantified as the number of flinches of the injected paw. Reduction of flinching was considered as antinociception. Ipsilateral, but not contralateral, peripheral administration of the 5-HT(1) receptor agonists R(+)-UH-301 (5-HT(1A); 0.1-3 microg/paw), CGS-12066A (5-HT(1B); 0.01-0.3 microg/paw), GR46611 (5-HT(1B/1D); 0.3-10 microg/paw), BRL54443 (5-HT(1E/1F); 3-300 microg/paw) or LY344864 (5-HT(1F); 3-300 microg/paw) significantly reduced formalin-induced flinching. The corresponding vehicle was devoid of any effect by itself. The local antinociceptive effect of R(+)-UH-301 (0.3 microg/paw) was significantly reduced by WAY-100635 (30-100 microg/paw; a 5-HT(1A) receptor antagonist). Moreover, the antagonists GR55562 (30-100 microg/paw; 5-HT(1B/D)) or SB224289 (30-100 microg/paw; 5-HT(1B)) dose-dependently reduced the antinociceptive effect of CGS-12066A (0.3 microg/paw) whereas GR55562 (30-100 microg/paw) or BRL15572 (30-100 microg/paw, 5-HT(1D)) reduced the antinociceptive effect of GR46611 (0.3 microg/paw). Interestingly, the effects of BRL54443 and LY344864 (300 microg/paw each) were partially reduced by methiothepin, but not by the highest doses of WAY-100635, SB224289 or BRL15572. The above antagonists did not produce any effect by themselves. These results suggest that peripheral activation of the 5-HT(1A,) 5-HT(1B), 5-HT(1D), 5-HT(1F) and, probably, 5-HT(1E) receptor subtypes leads to antinociception in the rat formalin test. Thus, the use of selective 5-HT(1) receptor agonists could be a therapeutic strategy to reduce inflammatory pain.