MilliporeSigma
  • Astrocyte elevated gene-1 regulates osteosarcoma cell invasion and chemoresistance via endothelin-1/endothelin A receptor signaling.
Astrocyte elevated gene-1 regulates osteosarcoma cell invasion and chemoresistance via endothelin-1/endothelin A receptor signaling.
Oncology letters (2013-02-20)
Bo Liu, Yi Wu, Dan Peng
ABSTRACT
Astrocyte elevated gene-1 (AEG-1) and endothelin-1 (ET-1)/endothelin A receptor (ETAR) signaling have been demonstrated to be important in osteosarcoma (OS) progression. In the present study, we explored the interaction between AEG-1 and ET-1/ETAR signaling in OS cells, and investigated the mechanism(s) through which the functional interaction may impact OS cell invasion and chemoresistance. Overexpression and knockdown of AEG-1 were performed in Saos-2 and MG-63 OS cells, respectively. Overexpression of AEG-1 in Saos-2 cells significantly increased ET-1 expression (at both the mRNA and protein levels), cell invasion, MMP-2 expression and cell survival against cisplatin. These effects were eradicated using a selective phosphatidylinositol 3-kinase (PI3K) inhibitor, LY294002, or a selective ETAR inhibitor, BQ123. Knockdown of AEG-1 in MG-63 cells significantly decreased ET-1 expression (at both the mRNA and protein levels), cell invasion, MMP-2 expression and cell survival against cisplatin. Exogenous ET-1 restored cell invasion and MMP-2 expression levels in MG-63 cells, in which AEG-1 had been knocked down, in the presence of LY294002, but not in the presence of BQ123. However, exogenous ET-1 only partially rescued cell survival against cisplatin-induced apoptosis in the presence of LY294002, in cells in which AEG-1 had been knocked down. In conclusion, we have demonstrated that AEG-1 regulates ET-1 expression at the transcriptional level in a PI3K-dependent manner in OS cells. Downstream of PI3K, ET-1/ETAR signaling primarily mediates the promoting effect of AEG-1 on OS cell invasion, likely through the upregulation of MMP-2 expression, thus, ET-1/ETAR signaling partially, but significantly, mediates the AEG-1-induced chemoresistance in OS cells. To the best of our knowledge, this study has provided the first evidence of a functional association between AEG-1 and ET-1/ETAR signaling in OS cells, which adds novel insights into the molecular mechanism of OS metastasis and chemoresistance.