MilliporeSigma
  • Expression of vitamin D receptor and metabolizing enzymes in multiple sclerosis-affected brain tissue.
Expression of vitamin D receptor and metabolizing enzymes in multiple sclerosis-affected brain tissue.
Journal of neuropathology and experimental neurology (2013-01-22)
Joost Smolders, Karianne G Schuurman, Miriam E van Strien, Jeroen Melief, Debbie Hendrickx, Elly M Hol, Corbert van Eden, Sabina Luchetti, Inge Huitinga
ABSTRACT
Vitamin D deficiency has been implicated as a risk factor for multiple sclerosis (MS), but how vitamin D metabolism affects MS pathophysiology is not understood. We studied the expression of vitamin D receptor (VDR) and related enzymes, including 1,25(OH)(2)D-24-hydroxylase (24-OHase; CYP24A1) and 25(OH)D-1α-hydroxylase (CYP27B1), in CNS tissues of 39 MS patients and 20 controls and in primary human glial cells in vitro. In control and MS normal-appearing white matter (NAWM), nuclear VDR immunostaining was observed in oligodendrocyte-like cells, human leukocyte antigen (HLA)-positive microglia, and glial fibrillary acidic protein-positive astrocytes. There was a 2-fold increase in VDR transcripts in MS NAWM versus control white matter (p = 0.03). In chronic active MS lesions, HLA-positive microglia/macrophages showed nuclear VDR staining; astrocytes showed nuclear and cytoplasmic VDR staining. Staining for 24-OHase was restricted to astrocytes.VDR and CYP27B1 mRNA expressions were increased in active MS lesions versus NAWM (p < 0.01, p = 0.04, respectively). In primary human astrocytes in vitro, the active form of vitamin D, 1,25(OH)(2)D(3), induced upregulation of VDR and CYP24A1. Tumor necrosis factor and interferon-γ upregulated CYP27B1 mRNA in primary human microglia and astrocytes. Increased VDR expression in MS NAWM and inflammatory cytokine-induced amplified expression of VDR and CYP27B1 in chronic active MS lesions suggest increased sensitivity to vitamin D in NAWM and a possible endogenous role for vitamin D metabolism in the suppression of active MS lesions.