MilliporeSigma
  • Effect of MNCQQ Cells on Migration of Human Dermal Fibroblast in Diabetic Condition.
Effect of MNCQQ Cells on Migration of Human Dermal Fibroblast in Diabetic Condition.
Biomedicines (2022-10-28)
Sen Jiang, Rie Ito-Hirano, Tsubame Nishikai-Yan Shen, Satoshi Fujimura, Hiroshi Mizuno, Rica Tanaka
ABSTRACT
A major symptom of diabetes mellitus (DM) is unfit hyperglycemia, which leads to impaired wound healing. It has been reported that the migration of fibroblasts can be suppressed under high glucose (HG) conditions. In our previous study, we introduced a serum-free culture method for mononuclear cells (MNCs) called quantity and quality control culture (QQc), which could improve the vasculogenic and tissue regeneration ability of MNCs. In this study, we described a culture model in which we applied a high glucose condition in human dermal fibroblasts to simulate the hyperglycemia condition in diabetic patients. MNC-QQ cells were cocultured with fibroblasts in this model to evaluate its role in improving fibroblasts dysfunction induced by HG and investigate its molecular mechanism. It was proven in this study that the impaired migration of fibroblasts induced by high glucose could be remarkably enhanced by coculture with MNC-QQ cells. PDGF B is known to play important roles in fibroblasts migration. Quantitative PCR revealed that MNC-QQ cells enhanced the gene expressions of PDGF B in fibroblasts under HG. Taken with these results, our data suggested a possibility that MNC-QQ cells accelerate wound healing via improving the fibroblasts migration and promote the gene expressions of PDGF B under diabetic conditions.