MilliporeSigma
The importance of natural IgM: scavenger, protector and regulator.
Nature reviews. Immunology (2010-10-16)
Michael R Ehrenstein, Clare A Notley
ABSTRACT
The existence of IgM has been known for more than a century, but its importance in immunity and autoimmunity continues to emerge. Studies of mice deficient in secreted IgM have provided unexpected insights into its role in several diverse processes, from B cell survival to atherosclerosis, as well as in autoimmunity and protection against infection. Among the various distinct properties that underlie the functions of IgM, two stand out: its polyreactivity and its ability to facilitate the removal of apoptotic cells. In addition, new B cell-targeted therapies for the treatment of autoimmunity have been shown to cause a reduction in serum IgM, potentially disrupting the functions of this immunoregulatory molecule and increasing susceptibility to infection.
Sigma-Aldrich
Anti-human IgM (10mg) KC, from mouse