MilliporeSigma
  • WIP1 Inhibition by GSK2830371 Potentiates HDM201 through Enhanced p53 Phosphorylation and Activation in Liver Adenocarcinoma Cells.
WIP1 Inhibition by GSK2830371 Potentiates HDM201 through Enhanced p53 Phosphorylation and Activation in Liver Adenocarcinoma Cells.
Cancers (2021-08-08)
Chiao-En Wu, Chen-Yang Huang, Chiao-Ping Chen, Yi-Ru Pan, John Wen-Cheng Chang, Jen-Shi Chen, Chun-Nan Yeh, John Lunec
ABSTRACT
Intrahepatic cholangiocarcinoma (iCCA) is an adenocarcinoma arising from the intrahepatic bile duct. It is the second most common primary liver cancer and has a poor prognosis. Activation of p53 by targeting its negative regulators, MDM2 and WIP1, is a potential therapy for wild-type p53 cancers, but few reports for iCCA or liver adenocarcinoma exist. Both RBE and SK-Hep-1 liver adenocarcinoma cell lines were treated with the HDM201 (Siremadlin) MDM2-p53 binding antagonist alone or in combination with the GSK2830371 WIP1 phosphatase inhibitor. Cell proliferation, clonogenicity, protein and mRNA expression, cell cycle distribution, and RNA sequencing were performed to investigate the effect and mechanism of this combination. GSK2830371 alone demonstrated minimal activity on proliferation and colony formation, but potentiated growth inhibition (two-fold decrease in GI50) and cytotoxicity (four-fold decrease in IC50) by HDM201 on RBE and SK-Hep-1 cells. HDM201 increased p53 protein expression, leading to transactivation of downstream targets (p21 and MDM2). Combination with GSK2830371 increased p53 phosphorylation, resulting in an increase in both p53 accumulation and p53-dependent trans-activation. G2/M arrest was observed by flow cytometry after this treatment combination. RNA sequencing identified 21 significantly up-regulated genes and five downregulated genes following p53 reactivation by HDM201 in combination with GSK2830371 at 6 h and 24 h time points compared with untreated controls. These genes were predominantly known transcriptional targets regulated by the p53 signaling pathway, indicating enhanced p53 activation as the predominant effect of this combination. The current study demonstrated that GSK2830371 enhanced the p53-dependent antiproliferative and cytotoxic effect of HDM201 on RBE and SK-Hep-1 cells, providing a novel strategy for potentiating the efficacy of targeting the p53 pathway in iCCA.
Sigma-Aldrich
Anti-MDM2 Antibody, clone 2A10, clone 2A10, from mouse