MilliporeSigma
  • Identification of a novel therapeutic candidate, NRK, in primary cancer-associated fibroblasts of lung adenocarcinoma microenvironment.
Identification of a novel therapeutic candidate, NRK, in primary cancer-associated fibroblasts of lung adenocarcinoma microenvironment.
Journal of cancer research and clinical oncology (2021-01-03)
Tongtong Wei, Jinjing Song, Kai Liang, Li Li, Xiaoxiang Mo, Zhiguang Huang, Gang Chen, Naiquan Mao, Jie Yang
ABSTRACT
Lung adenocarcinoma (LUAD) accounts for approximately half of patients in lung cancer. Cancer-associated fibroblasts (CAFs) are the major component in the tumor microenvironment (TME). Targeting CAFs is a promising therapeutic strategy for cancer treatment. However, therapeutic targets of CAFs in LUAD remains largely unclear. Seven CAFs and nine normal fibroblasts (NFs) were isolated from tumor and paratumor tissues of LUAD patients undergoing surgery, respectively. RNA-seq and bioinformatics analysis were performed to identify the differentially expressed genes (DEGs) and their functions in CAFs compared with NFs. DEGs of ten overlaying were obtained from RNA-seq, our previously reported lncRNA microarray and public datasets (E-MTAB-6149, E-MTAB-6653) and validated by RT-qPCR. Nik-related kinase (NRK) was further validated by RT-qPCR, immunofluorescence (IF), Western Blot (WB) in vitro, and in Cancer Cell Line Encyclopedia (CCLE) database. Survival analysis was performed on Kaplan-Meier plotter. A total of 1799 DEGs were identified, including 650 upregulated DEGs and 1149 downregulated DEGs. The upregulated and downregulated DEGs were mostly enriched in extracellular matrix (ECM) functions and in glycolysis/gluconeogenesis pathways. Interestingly, NRK was the most significantly upregulated overlaying DEGs which was rarely associated with CAFs before. NRK was predominantly expressed in CAFs, but weakly expressed in NFs, normal lung bronchial epithelial cell line BEAS-2B, LUAD cell lines A549 and H1299, as well as in the majority of 191 lung cancer cell lines including LUAD. Moreover, elevated NRK predicted poor survival in LUAD patients. Here, we first report that NRK is significantly elevated in LUAD-associated CAFs and may function as a promising therapeutic target for cancer combination treatment. Besides, modulation of ECM and glycolysis/gluconeogenesis pathways may be an efficient approach to alter CAFs functionality in LUAD.