MilliporeSigma
  • Eucalyptal A inhibits glioma by rectifying oncogenic splicing of MYO1B mRNA via suppressing SRSF1 expression.
Eucalyptal A inhibits glioma by rectifying oncogenic splicing of MYO1B mRNA via suppressing SRSF1 expression.
European journal of pharmacology (2020-10-26)
Dan Hua, Qian Zhao, Yang Yu, Huan Yu, Lin Yu, Xuexia Zhou, Qian Wang, Cuiyun Sun, Cuijuan Shi, Wenjun Luo, Zhendong Jiang, Weiting Wang, Lingli Wang, Dongli Zhang, Shengan Tang, Shizhu Yu
ABSTRACT
Glioma is the most common primary intracranial tumor, in which glioblastoma (GBM) is the most malignant and lethal. However, the current chemotherapy drugs are still unsatisfactory for GBM therapy. As the natural products mainly extracted from Eucalyptus species, phloroglucinol-terpene adducts have the potential to be anti-cancer lead compounds that attracted increasing attention. In order to discover the new lead compounds with the anti-GBM ability, we isolated Eucalyptal A with a phloroglucinol-terpene skeleton from the fruit of E. globulus and investigated its anti-GBM activity in vitro and in vivo. Functionally, we verified that Eucalyptal A could inhibit the proliferation, growth and invasiveness of GBM cells in vitro. Moreover, Eucalyptal A had the same anti-GBM activity in tumor-bearing mice as in vitro and prolonged the overall survival time by maintaining mice body weight. Further mechanism research revealed that Eucalyptal A downregulated SRSF1 expression and rectified SRSF1-guided abnormal alternative splicing of MYO1B mRNA, which led to anti-GBM activity through the PDK1/AKT/c-Myc and PAK/Cofilin axes. Taken together, we identified Eucalyptal A as an important anti-GBM lead compound, which represents a novel direction for glioma therapy.