MilliporeSigma
A reproduceable in situ xenograft model of spinal glioma.
Journal of neuroscience methods (2020-09-09)
Zhijian Weng, Liyi Ma, Junjie Li, Qiang Zhou, Lin Peng, Hong Li, Lei Chen, Zong Xin, Linyong Shi, Songtao Qi, Yuntao Lu
ABSTRACT
Spinal glioma is a nervous system tumor that tends to relapse and has no specific prognostic molecular biomarkers. Thus, a stable and reproduceable animal research model of spinal glioma is urgently needed. We established a new in situ tumor xenograft model of spinal glioma using nude mice. In this study, we implanted tumors into the cervical spinal cord of nude mice to mimic the pathological characteristics of the original tumors. Through anatomical experiments, we found that the cervical lamina of mice was thinner, the intervertebral space was much wider, and the adhesion muscles were more easily separated. According to the examination of spinal cord sections, the best puncture point we identified was located 0.9 mm lateral to the posterior median line at the level of the line between the midpoints of the scapulae and at a depth of 0.9 mm. In the nude mouse xenograft experiment, the implanted tumor tissue retained the pathological characteristics of the original tumor. This model used the cervical spinal cord as the puncture site and patient-derived primary tumor cells, which has never been performed before. Tumor cells could be injected directly without damaging the lamina. Thus, we could reduce the risk of man-made spinal cord injury and infection and avoid destroying the stability and integrity of the spine. This study established a stable and reliable animal model of spinal glioma for further molecular research and targeted therapy development.