MilliporeSigma
  • Highly efficient porous sorbent derived from asphalt for the solid-phase extraction of polycyclic aromatic hydrocarbons.
Highly efficient porous sorbent derived from asphalt for the solid-phase extraction of polycyclic aromatic hydrocarbons.
Journal of chromatography. A (2020-10-03)
Muhammad Kamran, Mohammed Dauda, Chanbasha Basheer, Mohammad Nahid Siddiqui, Hian Kee Lee
ABSTRACT
Polycyclic aromatic hydrocarbons (PAHs) are generated primarily during the incomplete combustion of organic matter and are ubiquitous environmental pollutants. For the first time, in this study, a mesoporous carbon derived from asphalt with high surface area (2300 m²g-1 with an average of 1.2 cm³ g-1) was utilized as a sorbent for the solid-phase extraction (SPE) of several PAHs in tap water samples. The factors influencing the extraction capability of the new material were investigated and the optimum conditions were determined to be as follows: Sample volume - 200 mL, no adjustment of sample pH, and sorbent amount - 50 mg. Under the most favorable SPE conditions, with gas chromatography-mass spectrometric analysis, the method exhibited a linear range of 0.5-50 μgL-1 with limits of detection between 0.004 and 0.026 μgL-1. The recoveries obtained from spiked tap water samples spiked at 1 μgL-1 and 5 μgL-1, were in the range 86.7-98.2% with relative standard deviations of <9%. The method was also applied to tap water samples collected from the local environment. The concentrations of PAHs detected ranged between 0.13 and 48 μgL-1. The reusability of the sorbent was tested with five consecutive SPE extraction, and no carryover of analytes was observed.