MilliporeSigma
  • WIP1 promotes cancer stem cell properties by inhibiting p38 MAPK in NSCLC.
WIP1 promotes cancer stem cell properties by inhibiting p38 MAPK in NSCLC.
Signal transduction and targeted therapy (2020-04-17)
Kaiyuan Deng, Liang Liu, Xiaoming Tan, Zhen Zhang, Jianjun Li, Yang Ou, Xin Wang, Shuang Yang, Rong Xiang, Peiqing Sun
ABSTRACT
Cancer stem cells (CSCs) are a small population of stem cell-like cancer cells that can initiate tumors in vivo, and are the major source of cancer initiation, relapse, and drug resistance. We previously reported that the p38 MAPK, through its downstream effectors MK2 and HSP27, suppressed CSC properties by downregulating the expression of transcription factors that mediate stemness in non-small-cell lung cancer (NSCLC) cells, and that despite unaltered total expression of total p38 proteins, the levels of activated p38 were reduced in NSCLC tissues. However, the mechanism underlying the reduced levels of activated p38 in NSCLC is unknown. In this study, we identified WIP1, a p38 phosphatase frequently overexpressed in cancer, as a suppressor of p38 in a pathway that regulates CSC properties in NSCLC. Increased WIP1 expression correlated with reduced levels of activated p38, and with increased levels of a CSC marker in NSCLC tissues. Further investigation revealed that WIP1 promoted stemness-related protein expression and CSC properties by inhibiting p38 activity in NSCLC cells. WIP1 inhibitors are currently under development as anticancer drugs based on their ability to reactivate p53. We found that a WIP1 inhibitor suppressed stemness-related protein expression and CSC properties by activating p38 in NSCLC cells in vitro and in vivo. These studies have identified the WIP1-p38-MK2-HSP27 cascade as a novel signaling pathway that, when altered, promotes CSC properties in NSCLC development, and have defined novel mechanisms underlying the oncogenic activity of WIP1 and the anticancer efficacy of WIP1 inhibitors.