MilliporeSigma
  • High methylation of lysine acetyltransferase 6B is associated with the Cobb angle in patients with congenital scoliosis.
High methylation of lysine acetyltransferase 6B is associated with the Cobb angle in patients with congenital scoliosis.
Journal of translational medicine (2020-05-26)
Yuantao Wu, Hongqi Zhang, Mingxing Tang, Chaofeng Guo, Ang Deng, Jiong Li, Yunjia Wang, Lige Xiao, Guanteng Yang
ABSTRACT
The etiology of congenital scoliosis (CS) is complex and uncertain. Abnormal DNA methylation affects the growth and development of spinal development. In this study, we investigated the role of DNA methylation in CS. The target region DNA methylation level in the peripheral blood of patients with CS was analyzed. Through in-depth analysis, genes closely related to the growth and development of the vertebra were identified. EdU staining was performed to verify the role of differentially expressed genes in chondrocyte proliferation. The hypermethylated KAT6B gene was observed in patients with CS, and was positively correlated with the Cobb angle. KAT6B was primarily expressed on chondrocytes. The promoter of KAT6B in CS patients was hypermethylated, and its expression was significantly reduced. Further mechanistic studies revealed that EZH2 mediated trimethylation of lysine 27 on histone H3 of the KAT6B promoter. Overexpression of KAT6B in CS-derived primary chondrocytes can significantly promote chondrocyte proliferation, which may be related to activation of the RUNX2/Wnt/β-catenin signaling pathway. Epigenetic modification of KAT6B may be a cause of CS. If similar epigenetic modification abnormalities can be detected through maternal liquid biopsy screening, they may provide useful biomarkers for early screening and diagnosis of CS.