MilliporeSigma
  • Optimized photo-stimulation of halorhodopsin for long-term neuronal inhibition.
Optimized photo-stimulation of halorhodopsin for long-term neuronal inhibition.
BMC biology (2019-11-30)
Chuanqiang Zhang, Shang Yang, Tom Flossmann, Shiqiang Gao, Otto W Witte, Georg Nagel, Knut Holthoff, Knut Kirmse
ABSTRACT
Optogenetic silencing techniques have expanded the causal understanding of the functions of diverse neuronal cell types in both the healthy and diseased brain. A widely used inhibitory optogenetic actuator is eNpHR3.0, an improved version of the light-driven chloride pump halorhodopsin derived from Natronomonas pharaonis. A major drawback of eNpHR3.0 is related to its pronounced inactivation on a time-scale of seconds, which renders it unsuited for applications that require long-lasting silencing. Using transgenic mice and Xenopus laevis oocytes expressing an eNpHR3.0-EYFP fusion protein, we here report optimized photo-stimulation techniques that profoundly increase the stability of eNpHR3.0-mediated currents during long-term photo-stimulation. We demonstrate that optimized photo-stimulation enables prolonged hyperpolarization and suppression of action potential discharge on a time-scale of minutes. Collectively, our findings extend the utility of eNpHR3.0 to the long-lasting inhibition of excitable cells, thus facilitating the optogenetic dissection of neural circuits.
Sigma-Aldrich
DNQX, ≥98% (TLC)