MilliporeSigma
  • Thymoquinone-PLGA-PVA Nanoparticles Ameliorate Bleomycin-Induced Pulmonary Fibrosis in Rats via Regulation of Inflammatory Cytokines and iNOS Signaling.
Thymoquinone-PLGA-PVA Nanoparticles Ameliorate Bleomycin-Induced Pulmonary Fibrosis in Rats via Regulation of Inflammatory Cytokines and iNOS Signaling.
Animals : an open access journal from MDPI (2019-11-14)
Sultan A M Saghir, Naif A Al-Gabri, Asmaa F Khafaga, Nahla H El-Shaer, Khaled A Alhumaidh, Mohamed F Elsadek, Badreldin M Ahmed, Daniyah M Alkhawtani, Mohamed E Abd El-Hack
ABSTRACT
Pulmonary fibrosis is considered one of the most chronic interstitial illnesses which are not easily treated. thymoquinone's (TQ) benefits are still partly problematic due to poor water solubility; therefore, it was loaded onto PLGA-PVA carriers. This study aimed to evaluate the potential effect of TQ-PLGA-PVA nanoparticles (TQ-PLGA-PVA-NPs) on pulmonary fibrosis induced by bleomycin in albino rats. Forty male rats were randomized into four groups. The first group served as the control group; the second and the third groups received bleomycin intratracheally, whereas the third group received TQ-PLGA-PVA-NPs after 4 weeks from bleomycin administration. The fourth group was administrated TQ-PLGA-PVA-NPs alone. The designed nanoparticles appeared around 20 nm size (10-30 nm), had a spherical shape, and had 80% encapsulation efficiency. The histological examination of rats simultaneously treated with TQ-PLGA-PVA-NPs and bleomycin revealed reduction in the thickness of the alveolar septa and improvement of the other lung structures, with the presence of lymphocytes admixed with exfoliated epithelium in a few lumina remaining. Ultrastructural findings revealed marked collagenolysis and the release of nanoparticles from ruptured pneumocytes within the alveolar septa after 14 days from TQ-PLGA-PVA-NPs administration. Very active pneumocyte types II were seen in the TQ-PLGA-PVANP group. Additionally, immunohistochemical expression of inducible nitric oxide (iNOS) and estimation of inflammatory cytokines in lung tissues including interleukin 10 (IL 10) and transforming growth factor-beta (TGF-β1) confirmed the antioxidant and anti-inflammatory effects of TQ-PLGA-PVANPs. The study concluded that TQ-PLGA-PVA-NPs could attenuate the bleomycin-induced pulmonary fibrosis, through the inhibition of lung inflammation and the suppression of bleomycin- induced oxidative stress.
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide (50:50), mol wt 30,000-60,000