MilliporeSigma
  • Immunomapping of desmosomal and nondesmosomal adhesion molecules in healthy canine footpad, haired skin and buccal mucosal epithelia: comparison with canine pemphigus foliaceus serum immunoglobulin G staining patterns.
Immunomapping of desmosomal and nondesmosomal adhesion molecules in healthy canine footpad, haired skin and buccal mucosal epithelia: comparison with canine pemphigus foliaceus serum immunoglobulin G staining patterns.
Veterinary dermatology (2010-08-27)
Petra Bizikova, Keith E Linder, Thierry Olivry
ABSTRACT
Pemphigus foliaceus (PF) is the most common canine autoimmune skin disease. In contrast to human PF (hPF), desmoglein-1 is a minor autoantigen in the canine disease. The major autoantigen(s) of canine PF (cPF) remain(s) unknown, which limits the ability to perform mechanistic studies of lesion formation and the development of novel diagnostic and therapeutic strategies for this disease. The immunofluorescence patterns of selected desmosomal (desmoglein-1, desmoglein-3, desmocollin-1, desmocollin-3, desmoplakin-1/2, plakoglobin and plakophilin-1) and nondesmosomal adhesion proteins (E-cadherin, claudin-1, zona occludens-1 and occludin) in healthy canine footpad, haired skin and buccal mucosal epithelia were determined using hPF and pemphigus vulgaris sera and specific antibodies. The immunostaining patterns were then compared with that of indirect immunofluorescence staining with 66 cPF sera. Most cPF sera (58 of 66; 88%) exhibited positive staining along keratinocyte margins in the stratum spinosum and stratum granulosum of canine footpad. One serum contained autoantibodies binding solely to stratum granulosum keratinocytes. Concurrent intercellular fluorescence in the stratum basale was limited to seven of 66 cPF sera (11%). Only 12 of 66 cPF sera (18%) also exhibited positive IF staining of the buccal mucosa. This study confirms the immunological heterogeneity of cPF immunoglobulin G autoantibodies. Moreover, the major indirect immunofluorescence staining pattern and the inability of most cPF sera to label the buccal mucosa closely matched that of desmocollin-1. These observations warrant further investigation of desmocollin-1 as a potential major cPF autoantigen.