MilliporeSigma
  • Plk1 protects kinetochore-centromere architecture against microtubule pulling forces.
Plk1 protects kinetochore-centromere architecture against microtubule pulling forces.
EMBO reports (2019-08-31)
Robert F Lera, Roshan X Norman, Marie Dumont, Alexandra Dennee, Joanne Martin-Koob, Daniele Fachinetti, Mark E Burkard
ABSTRACT
During mitosis, sister chromatids attach to microtubules which generate ~ 700 pN pulling force focused on the centromere. We report that chromatin-localized signals generated by Polo-like kinase 1 (Plk1) maintain the integrity of the kinetochore and centromere against this force. Without sufficient Plk1 activity, chromosomes become misaligned after normal condensation and congression. These chromosomes are silent to the mitotic checkpoint, and many lag and mis-segregate in anaphase. Their centromeres and kinetochores lack CENP-A, CENP-C, CENP-T, Hec1, Nuf2, and Knl1; however, CENP-B is retained. CENP-A loss occurs coincident with secondary misalignment and anaphase onset. This disruption occurs asymmetrically prior to anaphase and requires tension generated by microtubules. Mechanistically, centromeres highly recruit PICH DNA helicase and PICH depletion restores kinetochore disruption in pre-anaphase cells. Furthermore, anaphase defects are significantly reduced by tethering Plk1 to chromatin, including H2B, and INCENP, but not to CENP-A. Taken as a whole, this demonstrates that Plk1 signals are crucial for stabilizing centromeric architecture against tension.
Sigma-Aldrich
MISSION® esiRNA, targeting human ERCC6L