MilliporeSigma
  • Aldehyde dehydrogenase activity of breast cancer stem cells is primarily due to isoform ALDH1A3 and its expression is predictive of metastasis.
Aldehyde dehydrogenase activity of breast cancer stem cells is primarily due to isoform ALDH1A3 and its expression is predictive of metastasis.
Stem cells (Dayton, Ohio) (2011-02-01)
Paola Marcato, Cheryl A Dean, Da Pan, Rakhna Araslanova, Megan Gillis, Madalsa Joshi, Lucy Helyer, Luzhe Pan, Andrew Leidal, Shashi Gujar, Carman A Giacomantonio, Patrick W K Lee
ABSTRACT
Cancer stem cells (CSCs) are proposed to initiate cancer and propagate metastasis. Breast CSCs identified by aldehyde dehydrogenase (ALDH) activity are highly tumorigenic in xenograft models. However, in patient breast tumor immunohistological studies, where CSCs are identified by expression of ALDH isoform ALDH1A1, CSC prevalence is not correlative with metastasis, raising some doubt as to the role of CSCs in cancer. We characterized the expression of all 19 ALDH isoforms in patient breast tumor CSCs and breast cancer cell lines by total genome microarray expression analysis, immunofluorescence protein expression studies, and quantitative polymerase chain reaction. These studies revealed that ALDH activity of patient breast tumor CSCs and cell lines correlates best with expression of another isoform, ALDH1A3, not ALDH1A1. We performed shRNA knockdown experiments of the various ALDH isoforms and found that only ALDH1A3 knockdown uniformly reduced ALDH activity of breast cancer cells. Immunohistological studies with fixed patient breast tumor samples revealed that ALDH1A3 expression in patient breast tumors correlates significantly with tumor grade, metastasis, and cancer stage. Our results, therefore, identify ALDH1A3 as a novel CSC marker with potential clinical prognostic applicability, and demonstrate a clear correlation between CSC prevalence and the development of metastatic breast cancer.