MilliporeSigma
  • Occurrence of horizontal transmission in layer chickens after administration of an in ovo strain F Mycoplasma gallisepticum vaccine1,2,3.
Occurrence of horizontal transmission in layer chickens after administration of an in ovo strain F Mycoplasma gallisepticum vaccine1,2,3.
Poultry science (2019-06-11)
K E C Elliott, S L Branton, J D Evans, E D Peebles
ABSTRACT
In ovo vaccination is currently being considered as a means of delivery for live Mycoplasma gallisepticum (MG) vaccines. This study was performed to determine the transmissibility of strain F MG (FMG) from in ovo-vaccinated chicks to non-vaccinated pen mates. Eggs from an MG clean flock were incubated together for 18 D, at which point all live embryonated eggs were either not injected or administered a dilution of an FMG vaccine at 106 CFU per dose, 1 × 104 CFU per dose, 1 × 102 CFU per dose, or 1 CFU per dose. Non-injected eggs were hatched in a separate incubator. Ten non-injected, sentinel birds, and 1 in ovo-vaccinated FMG chick were placed in each of 32 isolation units located in 2 replicate rooms (8 replicates per dose). At 6 wk of age, surviving birds that had been vaccinated in ovo were removed, swabbed for FMG detection by PCR, and bled for serum plate agglutination (SPA) and ELISA testing for the presence of antibodies against MG (1, 2, 6, and all 8 in ovo-vaccinated chicks in the 106, 104, 102, and 1 CFU dosages). At 12 wk of age, the remaining sentinel birds were likewise sampled. No sentinel birds died. The in ovo-vaccinated birds that survived to 6 wk were serologically positive except for 5 birds in the 1 CFU treatment. The percentages of MG-positive sentinel birds and sentinel birds with antibody production against MG at 12 wk from each unit were not different between all MG dosages (P = 0.48, PCR; P = 0.77, SPA; P = 0.85, ELISA). Body weights of the in ovo-vaccinated chicks at 6 wk of age (P = 0.43) and the sentinel birds at 12 wk of age (P = 0.95) were each not affected by FMG treatment. These findings indicate that layer chickens in ovo vaccinated with a live-attenuated FMG vaccine were capable of transmitting FMG to other chicks with which they were in direct contact.