MilliporeSigma
  • Target of rapamycin in yeast, TOR2, is an essential phosphatidylinositol kinase homolog required for G1 progression.
Target of rapamycin in yeast, TOR2, is an essential phosphatidylinositol kinase homolog required for G1 progression.
Cell (1993-05-07)
J Kunz, R Henriquez, U Schneider, M Deuter-Reinhard, N R Movva, M N Hall
ABSTRACT
The yeast TOR2 gene encodes an essential 282 kd phosphatidylinositol (PI) 3-kinase homolog. TOR2 is related to the catalytic subunit of bovine PI 3-kinase and to yeast VPS34, a vacuolar sorting protein also shown to have PI 3-kinase activity. The immunosuppressant rapamycin most likely acts by inhibiting PI kinase activity because TOR2 mutations confer resistance to rapamycin and because a TOR1 TOR2 double disruption (TOR1 is a nonessential TOR2 homolog) confers G1 arrest, as does rapamycin. Our results further suggest that 3-phosphorylated phosphoinositides, whose physiological significance has not been determined, are an important signal in cell cycle activation. In yeast, this signal may act in a signal transduction pathway similar to the interleukin-2 signal transduction pathway in T cells.