MilliporeSigma
  • XPD suppresses cell proliferation and migration via miR-29a-3p-Mdm2/PDGF-B axis in HCC.
XPD suppresses cell proliferation and migration via miR-29a-3p-Mdm2/PDGF-B axis in HCC.
Cell & bioscience (2019-01-11)
Zhihua Xiao, Yijun Wang, Hao Ding
ABSTRACT
The aim of this study was to investigate the role of XPD in migration and invasion of hepatocellular carcinoma (HCC) cells. The expression of XPD and miR-29a-3p was examined by western blot and qRT-PCR, cell proliferation was detected by MTT assay, cell migration was detected by transwell assay. TargetScan was used to predict potential targets of miR-29a-3p. In this study, we found that the expression of XPD and miR-29a-3p was downregulated in HCC samples and HCC cell lines. XPD suppressed proliferation and migration of HCC cell via regulating miR-29a-3p expression. Target prediction analysis and dual-luciferase reporter assay confirmed Mdm2 and PDGF-B were direct targets of miR-29a-3p, and miR-29a-3p suppressed proliferation and migration of HCC cells via regulating the expression of Mdm2 or PDGF-B. Our data indicated that XPD suppressed cell proliferation and migration via miR-29a-3p-Mdm2/PDGF-B axis in HCC.