MilliporeSigma
  • Cell-traversal protein for ookinetes and sporozoites (CelTOS) formulated with potent TLR adjuvants induces high-affinity antibodies that inhibit Plasmodium falciparum infection in Anopheles stephensi.
Cell-traversal protein for ookinetes and sporozoites (CelTOS) formulated with potent TLR adjuvants induces high-affinity antibodies that inhibit Plasmodium falciparum infection in Anopheles stephensi.
Malaria journal (2019-04-25)
Sakineh Pirahmadi, Sedigheh Zakeri, Akram A Mehrizi, Navid D Djadid, Abbas-Ali Raz, Jafar J Sani, Ronak Abbasi, Zahra Ghorbanzadeh
ABSTRACT
Plasmodium falciparum parasite is the most deadly species of human malaria, and the development of an effective vaccine that prevents P. falciparum infection and transmission is a key target for malarial elimination and eradication programmes. P. falciparum cell-traversal protein for ookinetes and sporozoites (PfCelTOS) is an advanced vaccine candidate. A comparative study was performed to characterize the immune responses in BALB/c mouse immunized with Escherichia coli-expressed recombinant PfCelTOS (rPfCelTOS) in toll-like receptor (TLR)-based adjuvants, CpG and Poly I:C alone or in combination (CpG + Poly I:C), followed by the assessment of transmission-reducing activity (TRA) of anti-rPfCelTOS antibodies obtained from different vaccine groups in Anopheles stephensi. The aim of the current work was achieved by head-to-head comparison of the vaccine groups using conventional and avidity enzyme-linked immunosorbent assay (ELISA), immunofluorescence test (IFAT), and standard membrane feeding assay (SMFA). Comparing to rPfCelTOS alone, administration of rPfCelTOS with two distinct TLR-based adjuvants in vaccine mouse groups showed a significant increase in responses (antibody level, IgG subclass analysis, avidity, and Th1 cytokines) and was able to induce reasonable transmission-reducing activity. Also, comparable functional activity of anti-rPfCelTOS antibodies was found in group that received antigen in either CpG or Poly I:C (69.9%/20% and 73.5%/24.4%, respectively, reductions in intensity/prevalence). However, the vaccine group receiving rPfCelTOS in combination with CpG + Poly I:C showed a significant induction in antibody titers and inhibitory antibodies in oocysts development (78.3%/19.6% reductions in intensity/prevalence) in An. stephensi. A key finding in this investigation is that rPfCelTOS administered alone in BALB/c mouse is poorly immunogenic, with relatively low IgG level, avidity, inhibitory antibodies, and mixed Th1/Th2 responses. However, immunological characteristic (IgG level, cytophilic IgG2a and IgG2b, avidity, and Th1 cytokines) and TRA of anti-rPfCelTOS significantly enhanced in the presence of co-administration of TLR-based adjuvants, confirming that targeting TLRs would be an effective means for the enhancement of inducing TRA against rPfCelTOS.