MilliporeSigma
  • Levels and profiles of long-chain perfluoroalkyl carboxylic acids in Pacific cod from 14 sites in the North Pacific Ocean.
Levels and profiles of long-chain perfluoroalkyl carboxylic acids in Pacific cod from 14 sites in the North Pacific Ocean.
Environmental pollution (Barking, Essex : 1987) (2019-01-28)
Yukiko Fujii, Hayato Tuda, Yoshihisa Kato, Osamu Kimura, Tetsuya Endo, Kouji H Harada, Akio Koizumi, Koichi Haraguchi
ABSTRACT
We investigated the profiles and levels of perfluoroalkyl carboxylic acid (PFCA) contamination in Pacific cod (Gadus macrocephalus) from the North Pacific Ocean. The mean concentrations of PFCAs containing 8 to 14 carbon atoms (C8-C14) in edible Pacific cod muscle ranged from 216 to 670 pg g-1 wet weight in the Northeast Pacific Ocean (Seattle, Vancouver, Alaska, and Russia), from 819 to 1710 pg g-1 wet weight in Japanese coastal waters (Hokkaido, Aomori, Iwate, Miyagi, Tottori, and Shimane), and from 288 to 892 pg g-1 wet weight in Korean waters (Sokcho, Busan, and Yeosu). These results indicate there are geographical differences in the distribution of PFCAs. The long-chain PFCAs (C9-C14) contributed 96% of the total PFCA concentration across Japan, whereas they contributed only 33% of the total PFCA concentration in the USA and Canada. Long-chain PFCA concentrations in cod samples collected in Japanese and Korean waters were about three to four times those in samples from the USA, Canada, and Russia. Because seafood is considered an important dietary source of PFCAs, high concentrations of long-chain PFCAs in Pacific cod from Japanese and Korean waters may affect human dietary exposure and blood concentrations of long-chain PFCAs.