MilliporeSigma
  • More Than Black or White: Melanization and Toll Share Regulatory Serine Proteases in Drosophila.
More Than Black or White: Melanization and Toll Share Regulatory Serine Proteases in Drosophila.
Cell reports (2019-04-25)
Jan Paul Dudzic, Mark Austin Hanson, Igor Iatsenko, Shu Kondo, Bruno Lemaitre
ABSTRACT
The melanization response is an important defense mechanism in arthropods. This reaction is mediated by phenoloxidases (POs), which are activated by complex extracellular serine protease (SP) cascades. Here, we investigate the role of SPs in the melanization response using compound mutants in D. melanogaster and discover phenotypes previously concealed in single-mutant analyses. We find that two SPs, Hayan and Sp7, activate the melanization response in different manners: Hayan is required for blackening wound sites, whereas Sp7 regulates an alternate melanization reaction responsible for the clearance of Staphylococcus aureus. We present evidence that Sp7 is regulated by SPs activating the Toll NF-κB pathway, namely ModSP and Grass. Additionally, we reveal a role for the combined action of Hayan and Psh in propagating Toll signaling downstream of pattern recognition receptors activating either Toll signaling or the melanization response.