MilliporeSigma
  • Sphingosine-coating of plastic surfaces prevents ventilator-associated pneumonia.
Sphingosine-coating of plastic surfaces prevents ventilator-associated pneumonia.
Journal of molecular medicine (Berlin, Germany) (2019-06-22)
Aaron P Seitz, Fabian Schumacher, Jennifer Baker, Matthias Soddemann, Barbara Wilker, Charles C Caldwell, Ryan M Gobble, Markus Kamler, Katrin Anne Becker, Sascha Beck, Burkhard Kleuser, Michael J Edwards, Erich Gulbins
ABSTRACT
Ventilator-associated pneumonia (VAP) is a major cause of morbidity and mortality in critically ill patients. Here, we employed the broad antibacterial effects of sphingosine to prevent VAP by developing a novel method of coating surfaces of endotracheal tubes with sphingosine and sphingosine analogs. Sphingosine and phytosphingosine coatings of endotracheal tubes prevent adherence and mediate killing of Pseudomonas aeruginosa, Acinetobacter baumannii, and Staphylococcus aureus, even in biofilms. Most importantly, sphingosine-coating of endotracheal tubes also prevented P. aeruginosa and S. aureus pneumonia in vivo. Coating of the tubes with sphingosine was stable, without obvious side effects on tracheal epithelial cells and did not induce inflammation. In summary, we describe a novel method to coat plastic surfaces and provide evidence for the application of sphingosine and phytosphingosine as novel antimicrobial coatings to prevent bacterial adherence and induce killing of pathogens on the surface of endotracheal tubes with potential to prevent biofilm formation and VAP. KEY MESSAGES: Novel dip-coating method to coat plastic surfaces with lipids. Sphingosine and phytosphingosine as novel antimicrobial coatings on plastic surface. Sphingosine coatings of endotracheal tubes prevent bacterial adherence and biofilms. Sphingosine coatings of endotracheal tubes induce killing of pathogens. Sphingosine coatings of endotracheal tubes ventilator-associated pneumonia.