MilliporeSigma
Periostin concentrations in childhood-onset craniopharyngioma patients.
Journal of endocrinological investigation (2018-11-27)
K Heinks, C De Schutter-Nüsse, S Boekhoff, A Bogusz, J Zhu, J Peng, H L Müller
ABSTRACT
Periostin is highly expressed in craniopharyngioma (CP)-associated fibroblasts and has been identified as a marker for non-alcoholic fatty liver disease (NAFLD). Half of CP patients with hypothalamic syndrome develop NAFLD. We hypothesized that periostin concentration is elevated in biological fluids of CP and associated with pathological hepatic parameters, indicating increased risk for NAFLD. A cross-sectional study on 35 patients with sellar masses (SMP) recruited in the German Childhood Craniopharyngioma Registry (32 CP, 2 xanthogranuloma, 1 pilocytic astrocytoma), three short-statured patients with isolated growth hormone deficiency, five short-statured patients with normal findings in GH-stimulating tests and decreased insulin-like growth factor (IGF)-1 and seven healthy controls. Periostin was measured by Elisa in serum, urine and saliva. Periostin serum, urine and saliva concentrations in CP were similar to concentrations of the other groups. Hypothalamic involvement/hypothalamic lesions, degree of obesity as well as hepatic enzymes were not associated with elevated periostin concentrations. Due to low patient numbers with pathological hepatic parameters, missing imaging data on the degree of steatosis hepatis and the lack of histological proof of NAFLD, no definitive conclusions can be drawn from measured periostin concentrations in serum. Interestingly, the subgroup of patients with decreased IGF-1 levels showed elevated concentrations of serum periostin when compared with other groups. In CP, periostin concentrations are not associated with known risk factors for NAFLD such as hepatic and metabolic parameters, obesity and hypothalamic lesions. Accordingly, periostin does not seem to be a suitable marker for NAFLD in CP.