MilliporeSigma
  • Integrity of lichen cell membranes as an indicator of heavy-metal pollution levels in soil.
Integrity of lichen cell membranes as an indicator of heavy-metal pollution levels in soil.
Ecotoxicology and environmental safety (2019-03-01)
Piotr Osyczka, Kaja Rola
ABSTRACT
The epigeic lichens Cladonia rei and Diploschistes muscorum are effective heavy-metal-tolerant colonisers of highly polluted and disturbed sites. In this study we compare their bioaccumulation capacities, accumulation patterns, and responses to heavy-metal stress, as expressed in terms of cell membrane damage. We also aim at verifying the relationships between cell membrane damage and levels of soil pollution with heavy metals, and thereby to identify the bioindicative value of this physiological parameter. Total and intracellular concentrations of Zn, Pb, Cd, As, Cu, and Ni were measured in 140 samples of lichens and corresponding soil, collected from variously contaminated sites. Relative electrical conductivity (EC%) values were determined concurrently in the lichen samples. The studied lichens differ considerably in intracellular uptake susceptibility and the related reduction in membrane integrity. In C. rei thalli, more than half of Zn, Pb, Cd, and As loads are accumulated extracellularly, whereas D. muscorum exhibits a tendency towards intracellular accumulation of the same elements. This property is clearly reflected in cell membrane damage, which is considerably greater in the latter species irrespective of study site. This indicates that intracellular heavy-metal accumulation affects the level of cell membrane damage. Two soil pollution classes were distinguished for both lichens based on element contents in host-substrate samples. The losses of cell membrane integrity in lichen thalli are related to these classes. EC% values above 16 in C. rei and above 20 in D. muscorum suggest elevated levels of heavy metals in the soil. Consequently, this physiological parameter can serve as an early warning indicator for detection of elevated metal concentrations in soil. The biomonitoring method proposed here involves common and widespread lichen species and can be widely applied in post-industrial areas.